missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Europium(III) nitrate hexahydrate, 99.9% (REO), Alfa Aesar™, Reacton™

Manufacturer:  Alfa Aesar 1529004

 View more versions of this product

Catalog No. AA1529004



Specifications

Specifications

Europium(III) nitrate hexahydrate
99.9% (REO)
MFCD00149700
europium nitrate hexahydrate, europium iii nitrate, hexahydrate 1:3:6, nitric acid, europium 3+ salt, hexahydrate, eu.3no3.6h2o, ksc494m3l, europium 3+ trinitrate hexahydrate, europium iii nitrate hydrate, reacton, europium iii nitrate hexahydrate trace rare earth metals basis 1g
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Eu+3]
446.066
446.06 (265.90 Anhydrous)
Odorless
2g
10031-53-5
EuH12N3O15
Colorless crystalline
JVYYYCWKSSSCEI-UHFFFAOYSA-N
europium(3+);trinitrate;hexahydrate
202256
Crystalline
Colorless

Chemical Identifiers

10031-53-5
446.066
JVYYYCWKSSSCEI-UHFFFAOYSA-N
202256
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Eu+3]
EuH12N3O15
MFCD00149700
europium nitrate hexahydrate, europium iii nitrate, hexahydrate 1:3:6, nitric acid, europium 3+ salt, hexahydrate, eu.3no3.6h2o, ksc494m3l, europium 3+ trinitrate hexahydrate, europium iii nitrate hydrate, reacton, europium iii nitrate hexahydrate trace rare earth metals basis 1g
europium(3+);trinitrate;hexahydrate
Safety and Handling

Safety and Handling

H272-H315-H319-H335: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

EINECSNumber : 233-380-3

SDS Documents