Please login to your online account to display your discounted pricing

Europium(III) nitrate hexahydrate, 99.9% (REO), Alfa Aesar™, Reacton™

$103.35 - $265.19

Chemical Identifiers

Chemical Name or Material Europium(III) nitrate hexahydrate
CAS 10031-53-5
Physical Form Crystalline
Assay Percent Range 99.9% (REO)
Molecular Formula Eu(NO3)3·6H2O
Formula Weight 446.06; 265.90 (anhydrous)
MDL Number MFCD00149700
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1529004

  SDS
alfa aesar
1529004
2g Each for $103.35

AA1529018

  SDS
alfa aesar
1529018
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1529009

  SDS
alfa aesar
1529009
10g Each for $265.19
Description & Specifications

Specifications

Chemical Name or Material Europium(III) nitrate hexahydrate
CAS 10031-53-5
Health Hazard 1 H272-H315-H319-H335: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
EINECS Number 233-380-3
Physical Form Crystalline
Assay Percent Range 99.9% (REO)
Molecular Formula Eu(NO3)3·6H2O
Appearance Colorless crystalline
Formula Weight 446.06 (265.90 Anhydrous)
MDL Number MFCD00149700
Odor Odorless
Color Colorless
Quantity 2g
Safety and Handling

H272-H315-H319-H335: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.