Please login to your online account to display your discounted pricing

Alfa Aesar™ Europium(III) fluoride, anhydrous, REacton™, 99.98% (REO)

Chemical Identifiers

Chemical Name or Material Europium(III) fluoride
CAS 13765-25-8
Name Note Anhydrous
Purity Grade Reacton™
Formula Weight 208.96
MDL Number MFCD00010998
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1130204

  SDS
alfa aesar™
1130204
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1130206

  SDS
alfa aesar™
1130206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1130214

  SDS
alfa aesar™
1130214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 208.96
Name Note Anhydrous
Boiling Point >2280°C
TSCA TSCA
Quantity 2g
Health Hazard 1 Warning
MDL Number MFCD00010998
Melting Point 1390°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 99.98%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Purity Grade Reacton™
Chemical Name or Material Europium(III) fluoride
Sensitivity Hygroscopic
CAS 13765-25-8
Physical Form Powder
Assay Percent Notes (REO)
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.