missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Europium(III) fluoride, anhydrous, 99.5% (REO)

Manufacturer:  Alfa Aesar™ 8818103

 View more versions of this product

Catalog No. AA8818103



Specifications

Specifications

Europium (III) fluoride
13765-25-8
EuF3
europium fluoride, europium iii fluoride, acmc-20akvp, europium fluoride, europium 3+ ion trifluoride, europium iii fluoride, anhydrous trace rare earth metals basis 1g
Powder
−60 Mesh
>2280°C
1g
Anhydrous
(REO)
MFCD00010998
208.96
99.5%
Hygroscopic
1390°C

Chemical Identifiers

13765-25-8
EuF3
MFCD00010998
europium fluoride, europium iii fluoride, acmc-20akvp, europium fluoride, europium 3+ ion trifluoride, europium iii fluoride, anhydrous trace rare earth metals basis 1g
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents