Please login to your online account to display your discounted pricing

Europium(III) fluoride, 99+%, ACROS Organics™

$79.04 - $79.04

Chemical Identifiers

Chemical Name or Material Europium(III) fluoride
Name Note 99+%
CAS 13765-25-8
Assay Percent Range 99+%
Molecular Formula EuF3
Linear Formula EuF3
Formula Weight 208.96
MDL Number MFCD00010998
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC216860010

  SDS
thermo fisher scientific
216860010
1g Each for $79.04
Description & Specifications

Specifications

Formula Weight 208.96
Quantity 1g
EINECS Number 237-368-9
TSCA TSCA
Assay Percent Range 99+%
CAS 13765-25-8
Linear Formula EuF3
Molecular Formula EuF3
Chemical Name or Material Europium(III) fluoride
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 99+%
MDL Number MFCD00010998
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.