Please login to your online account to display your discounted pricing

Europium(III) chloride hydrate, Yellow crystalline, REacton™, 99.9% (REO), Alfa Aesar™

$108.64 - $108.64

Chemical Identifiers

Chemical Name or Material Europium(III) chloride hydrate
Physical Form Crystals
Assay Percent Range 99.9% (REO)
Molecular Formula EuCl3·xH2O
Formula Weight 258.32 (anhydrous)
MDL Number MFCD00010994
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1129806

  SDS
alfa aesar
1129806
5g Each for $108.64
Description & Specifications

Specifications

Formula Weight 258.32 (anhydrous)
Physical Form Crystals
Quantity 5g
Color Yellow
Odor Odorless
Molecular Formula EuCl3·xH2O
Chemical Name or Material Europium(III) chloride hydrate
Assay Percent Range 99.9% (REO)
MDL Number MFCD00010994