missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Europium(III) chloride hydrate, 99.99% (REO), Alfa Aesar™, REacton™

Manufacturer:  Alfa Aesar 1129706

 View more versions of this product

Catalog No. AA1129706



Specifications

Specifications

Europium(III) chloride hydrate
EuCl3·xH2O
Yellow crystalline
Crystalline
Yellow
99.99% (REO)
MFCD00010994
258.32 (anhydrous)
Odorless
5g

Chemical Identifiers

EuCl3·xH2O
MFCD00010994
Safety and Handling

Safety and Handling

EINECSNumber : 233-040-4

Documents