missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Europium(III) chloride hydrate, 99.99% (REO), Alfa Aesar™, REacton™

Chemical Identifiers

Physical Form Crystalline
Assay Percent Range 99.99% (REO)
Molecular Formula EuCl3·xH2O
Formula Weight 258.32 (Anhydrous)
MDL Number MFCD00010994
Chemical Name or Material Europium(III) chloride hydrate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1129703 SDS Alfa Aesar
1129703
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA1129706 SDS Alfa Aesar
1129706
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA1129714 SDS Alfa Aesar
1129714
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

Crystalline
EuCl3·xH2O
MFCD00010994
99.99% (REO)
258.32 (Anhydrous)
Europium(III) chloride hydrate

Specifications

Crystalline
EuCl3·xH2O
258.32 (Anhydrous)
Odorless
1g
99.99% (REO)
Yellow crystalline
MFCD00010994
Yellow
Europium(III) chloride hydrate

Safety and Handling

EINECSNumber : 233-040-4