missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Europium(III) chloride hexahydrate, 99.99%, (trace metal basis), Thermo Scientific™
Supplier: Thermo Scientific Chemicals 193010010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
for ultra trace analysisSpecifications
Europium(III) chloride hexahydrate | |
1g | |
Cl3Eu·6H2O | |
MFCD00149701 | |
10, 3854 | |
366.4 | |
99.99% | |
Glass bottle | |
4.8900g/mL |
13759-92-7 | |
39.0 to 42.5% (Eu) (Complexometry) | |
EuCl3·6H2O | |
15,186 | |
Solubility in water: soluble | |
366.4 | |
Trace Metal Basis | |
4.89 |
RUO â Research Use Only