missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Europium(III) carbonate hydrate, 99.99% (REO), Alfa Aesar™, REacton™
Manufacturer: Alfa Aesar 1321203
Specifications
Europium(III) carbonate hydrate | |
99.99% (REO) | |
MFCD00149702 | |
europium iii carbonate hydrate, 3co3.2eu.h2o, europium iii carbonate hydrate, reacton, dieuropium 3+ hydrate tricarbonate | |
C(=O)([O-])[O-].C(=O)([O-])[O-].C(=O)([O-])[O-].O.[Eu+3].[Eu+3] | |
501.967 | |
483.95 (Anhydrous) | |
Odorless | |
1g |
86546-99-8 | |
C3H2Eu2O10 | |
White powder | |
ZXWSECGKSXRERD-UHFFFAOYSA-H | |
europium(3+);tricarbonate;hydrate | |
91886282 | |
Powder | |
White |
Chemical Identifiers
86546-99-8 | |
501.967 | |
ZXWSECGKSXRERD-UHFFFAOYSA-H | |
91886282 | |
C(=O)([O-])[O-].C(=O)([O-])[O-].C(=O)([O-])[O-].O.[Eu+3].[Eu+3] |
C3H2Eu2O10 | |
MFCD00149702 | |
europium iii carbonate hydrate, 3co3.2eu.h2o, europium iii carbonate hydrate, reacton, dieuropium 3+ hydrate tricarbonate | |
europium(3+);tricarbonate;hydrate |
Safety and Handling
EINECSNumber : 227-582-0