missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Europium(III) acetate hydrate, REacton™, 99.999% (REO)

Manufacturer:  Alfa Aesar™ 4405618

 View more versions of this product

Catalog No. AA4405618



Specifications

Specifications

Europium (III) acetate hydrate
(REO)
MFCD00150115
RWVNKGWAXJFQSB-UHFFFAOYSA-N
acetic acid;europium;hydrate
16212055
Lumps/Powder
Reacton™
50g
62667-64-5
C6H14EuO7
bis acetyloxy europio acetate hydrate
CC(=O)O.CC(=O)O.CC(=O)O.O.[Eu]
350.135
329.10 (Anhydrous)
99.999%
Hygroscopic

Chemical Identifiers

62667-64-5
350.135
RWVNKGWAXJFQSB-UHFFFAOYSA-N
16212055
CC(=O)O.CC(=O)O.CC(=O)O.O.[Eu]
C6H14EuO7
MFCD00150115
bis acetyloxy europio acetate hydrate
acetic acid;europium;hydrate
Safety and Handling

Safety and Handling

EINECSNumber : 214-670-9

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents