Please login to your online account to display your discounted pricing

Alfa Aesar™ Europium(III) acetate hydrate, 99.99%

$235.51 - $235.51

Chemical Identifiers

Chemical Name or Material Europium(III) acetate hydrate
CAS 62667-64-5
Molecular Formula Eu(OOCCH3)3·xH2O
Formula Weight 329.10 (Anhydrous)
MDL Number MFCD00150115
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1806906

 
alfa aesar™
A1806906
5g Each for $235.51
Description & Specifications

Specifications

Formula Weight 329.10 (Anhydrous)
Chemical Name or Material Europium(III) acetate hydrate
TSCA TSCA
Assay 99.99%
Molecular Formula Eu(OOCCH3)3·xH2O
MDL Number MFCD00150115
CAS 62667-64-5
Quantity 5g