missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Europium(III) acetate hydrate, 99.99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1806903
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Europium(III) acetate hydrate | |
C6H9EuO6 | |
1 g | |
europium iii acetate hydrate 1:3:x, europium acetate hydrate, europium acetate,hydrate, europium iii acetatehydrate, c6h9euo6.h2o, europium iii acetate hydrate, reacton, europium iii acetate hydrate, reacton?, bis acetyloxy europio acetate hydrate, europium iii acetate hydrate trace metals basis | |
[Eu+3].CC([O-])=O.CC([O-])=O.CC([O-])=O | |
329.10 | |
329.10 (Anhydrous) |
62667-64-5 | |
MFCD00150115 | |
Hygroscopic | |
LNYNHRRKSYMFHF-UHFFFAOYSA-K | |
acetic acid;europium;hydrate | |
16212055 | |
99.99% |
Chemical Identifiers
62667-64-5 | |
329.10 | |
LNYNHRRKSYMFHF-UHFFFAOYSA-K | |
16212055 |
C6H9EuO6 | |
MFCD00150115 | |
europium iii acetate hydrate 1:3:x, europium acetate hydrate, europium acetate,hydrate, europium iii acetatehydrate, c6h9euo6.h2o, europium iii acetate hydrate, reacton, europium iii acetate hydrate, reacton?, bis acetyloxy europio acetate hydrate, europium iii acetate hydrate trace metals basis | |
[Eu+3].CC([O-])=O.CC([O-])=O.CC([O-])=O |
Safety and Handling
EINECSNumber : 214-670-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only