Please login to your online account to display your discounted pricing

Europium(III) acetate hydrate, 99.9% (REO), Alfa Aesar™ Reacton™

$131.46 - $503.34

Chemical Identifiers

Chemical Name or Material Europium(III) acetate hydrate
CAS 62667-64-5
Physical Form Crystalline Powder
Assay Percent Range 99.9% (REO)
Molecular Formula Eu(OOCCH3)3·xH2O
Formula Weight 329.10 (anhydrous)
MDL Number MFCD00150115
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4287806

 
alfa aesar
4287806
5g Each for $131.46

AA4287814

 
alfa aesar
4287814
25g Each for $503.34
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 329.10 (Anhydrous)
Quantity 5g
Color White
Chemical Name or Material Europium(III) acetate hydrate
EINECS Number 214-670-9
Assay Percent Range 99.9% (REO)
Molecular Formula Eu(OOCCH3)3·xH2O
Appearance White crystalline powder
MDL Number MFCD00150115
CAS 62667-64-5