missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Europium(III) 2,4-pentanedionate hydrate, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 1320909

 View more versions of this product

Catalog No. AA1320909



Specifications

Specifications

Europium(III) 2,4-pentanedionate hydrate
140°C (decomposition)
181266-82-0
C15H23EuO7
europium iii 2,4-pentanedionate hydrate reo
GLDOQTGHEKXAIO-KJVLTGTBSA-K
europium(3+);(Z)-4-oxopent-2-en-2-olate;hydrate
131675702
Powder
Hygroscopic
10g
(REO)
MFCD00192170
Insoluble in water.
CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].O.[Eu+3]
467.306
449.29 (Anhydrous)
99.9%

Chemical Identifiers

181266-82-0
467.306
GLDOQTGHEKXAIO-KJVLTGTBSA-K
131675702
CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].O.[Eu+3]
C15H23EuO7
MFCD00192170
europium iii 2,4-pentanedionate hydrate reo
europium(3+);(Z)-4-oxopent-2-en-2-olate;hydrate
Safety and Handling

Safety and Handling

EINECSNumber : 238-185-7

TSCA : No

Recommended Storage : Ambient temperatures

Documents