missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Europium(III) 2,4-pentanedionate hydrate, 99.9% (REO), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01320906
Description
It is used in various catalysts and catalytic reagents for organic synthesis, including the fabrication of various shapes of carbon nanostructures.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Europium(III) 2,4-pentanedionate hydrate | |
140°C (decomposition) | |
C15H21EuO6 | |
5 g | |
europium iii 2,4-pentanedionate hydrate reo | |
YMTKZYSHNNXTIJ-UHFFFAOYSA-N | |
europium(3+) tris(2,4-dioxopentan-3-ide) | |
131675702 | |
99.9% |
181266-82-0 | |
(REO) | |
MFCD00192170 | |
Hygroscopic | |
Insoluble in water. | |
[Eu+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O | |
449.29 | |
449.29 (Anhydrous) | |
Powder |
Chemical Identifiers
181266-82-0 | |
449.29 | |
YMTKZYSHNNXTIJ-UHFFFAOYSA-N | |
131675702 |
C15H21EuO6 | |
MFCD00192170 | |
europium iii 2,4-pentanedionate hydrate reo | |
[Eu+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O |
Safety and Handling
EINECSNumber : 238-185-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only