Please login to your online account to display your discounted pricing

Europium(II) bromide, White crystalline powder, 99.99% (Metals basis), Alfa Aesar™

$394.77 - $394.77

Chemical Identifiers

Chemical Name or Material Europium(II) bromide
CAS 13780-48-8
Physical Form Crystalline Powder
Assay Percent Range 99.99% (Metals basis)
Molecular Formula EuBr2
Formula Weight 311.77
MDL Number MFCD01076548
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4443103

  SDS
alfa aesar
4443103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4443114

  SDS
alfa aesar
4443114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4443106

  SDS
alfa aesar
4443106
5g Each for $394.77
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 311.77
Quantity 1g
Color White
Chemical Name or Material Europium(II) bromide
EINECS Number 13780-48-8
Odor Odorless
Assay Percent Range 99.99% (Metals basis)
Molecular Formula EuBr2
MDL Number MFCD01076548
CAS 13780-48-8
Health Hazard 1 H315-H319: Causes skin irritation. Causes serious eye irritation.
Health Hazard 2 In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
Safety and Handling

H315-H319: Causes skin irritation. Causes serious eye irritation.

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.