missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Europium ICP Standard, 1mL = 1mg Eu (1,000ppm Eu), Ricca Chemical

Manufacturer:  Ricca Chemical Company RIPEU1KN500

 View more versions of this product

Catalog No. PEU1KN-500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Europium ICP Standard
93.0,4.56,0.11
Eu2O3
[O-2].[O-2].[O-2].[Eu+3].[Eu+3]
351.925
Liquid
1,000ppm Eu
Colorless
1308-96-9,7732-18-5,1308-96-9
97.0,4.74,0.11
AEBZCFFCDTZXHP-UHFFFAOYSA-N
europium(3+);oxygen(2-)
159371
Poly natural
1 mL = 1 mg Eu
500mL

Chemical Identifiers

1308-96-9
351.925
159371
[O-2].[O-2].[O-2].[Eu+3].[Eu+3]
Eu2O3
AEBZCFFCDTZXHP-UHFFFAOYSA-N
europium(3+);oxygen(2-)
Documents