Please login to your online account to display your discounted pricing

Alfa Aesar™ Europium foil, 1.0mm (0.04 in.) thick, 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Europium foil
CAS 7440-53-1
Name Note 1.0mm (0.04 in.) thick
MDL Number MFCD00010992
UN Number UN3208
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA10174FF

  SDS
alfa aesar™
10174FF
25x25mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 25x25mm
Chemical Name or Material Europium foil
Odor Odorless
Packaging Mylar
TSCA TSCA
Name Note 1.0mm (0.04 in.) thick
Physical Form Foil
Assay 99.9%
Assay Percent Notes (REO)
MDL Number MFCD00010992
CAS 7440-53-1
UN Number UN3208
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H250-H261
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gas.
Health Hazard 3 GHS P Statement
P210-P222-P231+P232-P370+P378b-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not allow contact with air.
Handle under inert gas.
Protect from moisture.
In case of fire: Use for
Storage Note 1
  • Packaged under argon
DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: METALLIC SUBSTANCE, WATER-REACTIVE, N.O.S.
Weight ≈3.3g/25x25mm

  • Packaged under argon

Safety and Handling

Danger

GHS P Statement
P210-P222-P231+P232-P370+P378b-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not allow contact with air.
Handle under inert gas.
Protect from moisture.
In case of fire: Use for

GHS H Statement
H250-H261
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gas.

  • Packaged under argon