missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Europium foil, 0.25mm (0.01in) thick, 99.9% (REO), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 010175FI

 View more versions of this product

Catalog No. AA10175FI


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Specifications

Gray
0.25mm (0.01 in.) thick
(REO)
MFCD00010992
50 x 50 mm
OGPBJKLSAFTDLK-UHFFFAOYSA-N
europium
23981
99.9%
Mylar under Argon
Foil
Europium foil
7440-53-1
Eu
UN3208
europio, atom, ingot, acmc-1bj8v, europium, chip, in mineral oil, foil, 1.0mm 0.04in thick, foil, 0.1mm 0.004in thick, foil, 0.25mm 0.01in thick, rod, 9.52 mm 0.375in dia, ingot reo , packed in mineral oil
[Eu]
151.96
CHEBI:32999
Odorless
≈0.82g/25x25mm

Chemical Identifiers

7440-53-1
151.96
OGPBJKLSAFTDLK-UHFFFAOYSA-N
23981
europium
Eu
MFCD00010992
europio, atom, ingot, acmc-1bj8v, europium, chip, in mineral oil, foil, 1.0mm 0.04in thick, foil, 0.1mm 0.004in thick, foil, 0.25mm 0.01in thick, rod, 9.52 mm 0.375in dia, ingot reo , packed in mineral oil
CHEBI:32999
[Eu]
Safety and Handling

Safety and Handling

GHS H Statement
H250-H261

P210-P222-P223-P231+P232-P280-P335+P334-P370+P378q-P501c

H250-H261-H500

DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: METALLIC SUBSTANCE, WATER-REACTIVE, N.O.S.

EINECSNumber : 231-161-7

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only