Learn More
Eugenol, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 119111000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Eugenol | |
97-53-0 | |
Yellow | |
254.0°C | |
Authentic | |
Glass bottle | |
1.5400 to 1.5420 | |
MFCD00008654 | |
06,961 | |
15,394 | |
Solubility in water: 2.46g/L (25°C). Other solubilities: soluble in glacial acetic acid,soluble in alcohol,chloroform and ether | |
COC1=CC(CC=C)=CC=C1O | |
164.20 | |
CHEBI:4917 | |
99% |
99% | |
-12°C to 10°C | |
1.0600g/mL | |
>112°C | |
98.5% min. (GC) | |
C10H12O2 | |
(H2C=CHCH2)C6H3(OCH3)OH | |
100 g | |
1.06 | |
eugenol, 4-allylguaiacol, 4-allyl-2-methoxyphenol, eugenic acid, allylguaiacol, caryophyllic acid, p-allylguaiacol, p-eugenol, 2-methoxy-4-allylphenol, engenol | |
RRAFCDWBNXTKKO-UHFFFAOYSA-N | |
2-methoxy-4-prop-2-enylphenol | |
3314 | |
164.2 | |
Liquid |
Chemical Identifiers
97-53-0 | |
164.20 | |
RRAFCDWBNXTKKO-UHFFFAOYSA-N | |
3314 | |
COC1=CC(CC=C)=CC=C1O |
C10H12O2 | |
MFCD00008654 | |
eugenol, 4-allylguaiacol, 4-allyl-2-methoxyphenol, eugenic acid, allylguaiacol, caryophyllic acid, p-allylguaiacol, p-eugenol, 2-methoxy-4-allylphenol, engenol | |
CHEBI:4917 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice
GHS Signal Word: Warning
EINECSNumber : 202-589-1
RTECSNumber : SJ4375000
TSCA : TSCA
RUO – Research Use Only