Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Ets-1 Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Ets-1 Ab-1, Mouse Monoclonal Antibody.

$524.59 - $1,536.51

Specifications

Antigen Ets-1 Ab-1
Clone 1G11
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MS1762R7

 
thermo scientific™ lab vision™
MS1762R7
7mL Each for $968.20

MS1762S

 
thermo scientific™ lab vision™
MS1762S
1mL Each for $1,536.51

MS1762S0

 
thermo scientific™ lab vision™
MS1762S0
100µL Each for $524.59

MS1762S1

 
thermo scientific™ lab vision™
MS1762S1
500µL Each for $1,200.08
Description & Specifications

Specifications

Antigen Ets-1 Ab-1
Clone 1G11
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Mouse
Regulatory Status RUO
Research Discipline Transcription Regulation
Type Primary
Immunogen Recombinant oncoprotein corresponding to aa 226-454 of human Ets-1
Isotype IgG2a kappa

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Ets-1 Ab-1, Mouse Monoclonal Antibody.

Ets-1 is a transcription factor involved in the regulation of genes required for extracellular matrix remodeling during metastasis of tumors. It is known to interact with the enhancer of urokinase like plasminogen activator, and promoters of stromelysin-1 and collagenase-1. It is absent from normal gastric mucosa but is expressed in 60% of gastric carcinoma and oral squamous cell carcinoma.

Host Species: Mouse

Clone: 1G11

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: aa 226-454

Immunogen: Recombinant oncoprotein corresponding to aa 226-454 of human Ets-1

Molecular Weight: 51 kDa

Positive Control: Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO