missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Etoposide, TCI America™

Manufacturer:  TCI America E06751G

 View more versions of this product

Catalog No. E06751G



Specifications

Specifications

Etoposide
251°C
33419-42-0
vjjpusntgommgy-nzlmilqcsa
CC1OCC2C(O1)C(C(C(O2)OC3C4COC(=O)C4C(C5=CC6=C(C=C35)OCO6)C7=CC(=C(C(=C7)OC)O)OC)O)O
588.562
588.56
≥98.0% (HPLC)
White
1g
C29H32O13
VJJPUSNTGOMMGY-NZLMILQCSA-N
(9R)-5-[[(2R,6R,7R,8R)-7,8-dihydroxy-2-methyl-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-6-yl]oxy]-9-(4-hydroxy-3,5-dimethoxyphenyl)-5a,6,8a,9-tetrahydro-5H-[2]benzofuro[6,5-f][1,3]benzodioxol-8-one
50936917
Crystal-Powder at 20°C

Chemical Identifiers

33419-42-0
588.562
vjjpusntgommgy-nzlmilqcsa
(9R)-5-[[(2R,6R,7R,8R)-7,8-dihydroxy-2-methyl-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-6-yl]oxy]-9-(4-hydroxy-3,5-dimethoxyphenyl)-5a,6,8a,9-tetrahydro-5H-[2]benzofuro[6,5-f][1,3]benzodioxol-8-one
C29H32O13
VJJPUSNTGOMMGY-NZLMILQCSA-N
50936917
CC1OCC2C(O1)C(C(C(O2)OC3C4COC(=O)C4C(C5=CC6=C(C=C35)OCO6)C7=CC(=C(C(=C7)OC)O)OC)O)O
Safety and Handling

Safety and Handling

RTECSNumber : KC0190000

TSCA : No

Documents