Please login to your online account to display your discounted pricing

Ethynylmagnesium chloride, 0.6M solution in THF/Toluene, AcroSeal™, ACROS Organics™


Quantity 800mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC388958000

 View more versions of this product

Catalog No. AC388958000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Health Hazard 3 GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Health Hazard 2 GHS H Statement
Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. Causes severe skin burns and eye damage. May cause respiratory irritation. Suspected of damaging the unborn child. Reacts violently with water. May form explosive peroxides. Suspected of causing cancer. May be fatal if swallowed and enters airways. May cause damage to organs through prolonged or repeated exposure if inhaled. May cause drowsiness or dizziness.
Formula Weight 84.79
Physical Form Liquid
Quantity 800mL
Color Clear brown to dark brown
Packaging AcroSeal™ Glass Bottle
Density 0.9600g/mL
Linear Formula HCidentical toCMgCl
CAS 108-88-3
Storage Information Product tends to crystallize at lower temperatures. Store above 15°C. Precipitates can be redissolved by warming and appropriate homogenization.
Chemical Name or Material Ethynylmagnesium chloride
Name Note 0.6M solution in THF/Toluene
CAS 65032-27-1
Additional Information contains about 4 to 7% toluene
Concentration 4 to 6%
CAS 109-99-9
Health Hazard 1 Danger
MDL Number MFCD00075343
Molecular Formula C2HClMg

Specifications

Chemical Name or Material Ethynylmagnesium chloride
Name Note 0.6M solution in THF/Toluene
CAS 65032-27-1
CAS 108-88-3
CAS 109-99-9
Molecular Formula C2HClMg
Linear Formula HCidentical toCMgCl
Formula Weight 84.79
MDL Number MFCD00075343
Accessories

Danger

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. Causes severe skin burns and eye damage. May cause respiratory irritation. Suspected of damaging the unborn child. Reacts violently with water. May form explosive peroxides. Suspected of causing cancer. May be fatal if swallowed and enters airways. May cause damage to organs through prolonged or repeated exposure if inhaled. May cause drowsiness or dizziness.

SDS & Safety Documents