Please login to your online account to display your discounted pricing

Ethynylmagnesium chloride, 0.6M solution in THF/Toluene, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle in Tin Can

Manufacturer: thermo fisher scientific  AC388951000

 View more versions of this product

Catalog No. AC388951000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 84.79
Quantity 100mL
Color Clear brown to dark brown
Packaging AcroSeal™ Glass Bottle in Tin Can
Density 0.9600g/mL
Concentration 4 to 6%
Linear Formula HCidentical toCMgCl
Additional Information contains about 4 to 7% toluene
CAS 109-99-9
Chemical Name or Material Ethynylmagnesium chloride
Name Note 0.6M solution in THF/Toluene
Molecular Formula C2HClMg
MDL Number MFCD00075343
CAS 65032-27-1
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. Causes severe skin burns and eye damage. May cause respiratory irritation. Suspected of
CAS 108-88-3
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Do not breathe dust/fume/gas/mist/vapors/spray. Protect from moisture. IF IN E
Storage Information Product tends to crystallize at lower temperatures. Store above 15°C. Precipitates can be redissolved by warming and appropriate homogenization.

Specifications

Chemical Name or Material Ethynylmagnesium chloride
Name Note 0.6M solution in THF/Toluene
CAS 65032-27-1
CAS 108-88-3
CAS 109-99-9
Molecular Formula C2HClMg
Linear Formula HCidentical toCMgCl
Formula Weight 84.79
MDL Number MFCD00075343
Accessories

Danger

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Do not breathe dust/fume/gas/mist/vapors/spray. Protect from moisture. IF IN E

GHS P Statement Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. Causes severe skin burns and eye damage. May cause respiratory irritation. Suspected of

SDS & Safety Documents