Learn More
Ethynylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific AC439121000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Ethynylmagnesium bromide | |
Colorless to Gray | |
C2HBrMg | |
−17°C | |
LROBJRRFCPYLIT-UHFFFAOYSA-M | |
magnesium;ethyne;bromide | |
4071243 | |
Liquid | |
0.94 | |
0.40 to 0.60M |
4301-14-8,109-99-9 | |
100mL | |
CHCMgBr | |
bromo ethynyl magnesium, ethinyl magnesium bromide, ethinylmagnesium bromide, ethynyl magnesium bromide, ethynyl magnesiumbromide, ethynyl-magnesium bromide, ethynylmagnesium bromide, ethynylmagnesium bromide solution, grignard reagent, hugjuypsxulvqq-uhfffaoysa-m, pubchem18180 | |
C#[C-].[Mg+2].[Br-] | |
129.24 | |
129.24 | |
AcroSeal™ Glass Bottle | |
May develop some turbidity | |
0.9400g/mL |
Chemical Identifiers
4301-14-8 | |
129.24 | |
bromo ethynyl magnesium, ethinyl magnesium bromide, ethinylmagnesium bromide, ethynyl magnesium bromide, ethynyl magnesiumbromide, ethynyl-magnesium bromide, ethynylmagnesium bromide, ethynylmagnesium bromide solution, grignard reagent, hugjuypsxulvqq-uhfffaoysa-m, pubchem18180 | |
magnesium;ethyne;bromide |
C2HBrMg | |
LROBJRRFCPYLIT-UHFFFAOYSA-M | |
4071243 | |
C#[C-].[Mg+2].[Br-] |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Reacts violently w
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture
GHS Signal Word: Danger
RUO – Research Use Only