missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethynylbenzene, TCI America™

Manufacturer:  TCI America E0196500ML

 View more versions of this product

Catalog No. E0196500ML



Specifications

Specifications

Ethynylbenzene
Yellow
536-74-3
MFCD00008570
phenylacetylene, phenylethyne, benzene, ethynyl, 1-phenylethyne, phenylacetylide, phenyl acetylene, ethyne, phenyl, acetylene, phenyl, ethynyl-benzene, phenylacethylene
C#CC1=CC=CC=C1
102.136
102.14
≥98.0% (GC)
142°C
500mL
C8H6
3295
UEXCJVNBTNXOEH-UHFFFAOYSA-N
ethynylbenzene
10821
Clear Liquid at 20°C

Chemical Identifiers

536-74-3
102.136
UEXCJVNBTNXOEH-UHFFFAOYSA-N
10821
C#CC1=CC=CC=C1
C8H6
MFCD00008570
phenylacetylene, phenylethyne, benzene, ethynyl, 1-phenylethyne, phenylacetylide, phenyl acetylene, ethyne, phenyl, acetylene, phenyl, ethynyl-benzene, phenylacethylene
ethynylbenzene
Safety and Handling

Safety and Handling

RTECSNumber : DA0780000

TSCA : Yes

Recommended Storage : Refrigerator

Documents