missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethynyl p-Tolyl Sulfone, TCI America™

Manufacturer:  TCI America E04665G

 View more versions of this product

Catalog No. E04665G



Specifications

Specifications

Ethynyl p-Tolyl Sulfone
5g
C9H8O2S
p-Toluenesulfonylacetylene, Tosylacetylene
CC1=CC=C(C=C1)S(=O)(=O)C#C
180.221
180.22
≥98.0% (GC)
White-Yellow
13894-21-8
MFCD00191647
FTHKWIMQNXVEHW-UHFFFAOYSA-N
1-ethynylsulfonyl-4-methylbenzene
4463273
Crystal-Powder at 20°C

Chemical Identifiers

13894-21-8
180.221
FTHKWIMQNXVEHW-UHFFFAOYSA-N
4463273
CC1=CC=C(C=C1)S(=O)(=O)C#C
C9H8O2S
MFCD00191647
p-Toluenesulfonylacetylene, Tosylacetylene
1-ethynylsulfonyl-4-methylbenzene
Safety and Handling

Safety and Handling

TSCA : No

Documents