Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyltriphenylphosphonium chloride, 98%

Chemical Identifiers

Chemical Name or Material Ethyltriphenylphosphonium chloride
CAS 896-33-3
Molecular Formula CH3CH2P(C6H5)3Cl
Formula Weight 326.81
MDL Number MFCD00044699
UN Number UN3077
Beilstein 4170030
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAB2301514

  SDS
alfa aesar™
B2301514
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2301522

  SDS
alfa aesar™
B2301522
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2301536

  SDS
alfa aesar™
B2301536
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 326.81
Molecular Formula CH3CH2P(C6H5)3Cl
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 25g
UN Number UN3077
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA TSCA
Beilstein 4170030
Health Hazard 1 Danger
Assay 98%
CAS 896-33-3
Health Hazard 2 GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point ≈235°C (dec.)
Chemical Name or Material Ethyltriphenylphosphonium chloride
MDL Number MFCD00044699
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.