Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyltrimethoxysilane, 97%

Chemical Identifiers

Chemical Name or Material Ethyltrimethoxysilane
CAS 5314-55-6
Molecular Formula (CH3O)3SiCH2CH3
Formula Weight 150.25
MDL Number MFCD00051541
UN Number UN1993
Synonym Trimethoxyethylsilane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2275214

  SDS
alfa aesar™
B2275214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2275222

  SDS
alfa aesar™
B2275222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 150.25
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
CAS 5314-55-6
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Ethyltrimethoxysilane
UN Number UN1993
Quantity 25g
Health Hazard 1 Warning
Boiling Point 123° to 124°C
Molecular Formula (CH3O)3SiCH2CH3
Synonym Trimethoxyethylsilane
Flash Point 27°C (81°F)
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
TSCA TSCA
MDL Number MFCD00051541
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.