missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyltrichlorosilane, TCI America™

Manufacturer:  TCI America E0188500G

 View more versions of this product

Catalog No. E0188500G



Specifications

Specifications

Ethyltrichlorosilane
Yellow
115-21-9
MFCD00000485
ethyltrichlorosilane, trichloro ethyl silane, trichloroethylsilicane, silane, trichloroethyl, ethyl silicon trichloride, trichloroethylsilicon, silicane, trichloroethyl, ethyl trichlorosilane, silane, ethyltrichloro, c2h5sicl3
CC[Si](Cl)(Cl)Cl
163.497
163.50
≥98.0% (GC,T)
100°C
500g
C2H5Cl3Si
1196
ZOYFEXPFPVDYIS-UHFFFAOYSA-N
trichloro(ethyl)silane
8260
Clear Liquid at 20°C

Chemical Identifiers

115-21-9
163.497
ZOYFEXPFPVDYIS-UHFFFAOYSA-N
8260
CC[Si](Cl)(Cl)Cl
C2H5Cl3Si
MFCD00000485
ethyltrichlorosilane, trichloro ethyl silane, trichloroethylsilicane, silane, trichloroethyl, ethyl silicon trichloride, trichloroethylsilicon, silicane, trichloroethyl, ethyl trichlorosilane, silane, ethyltrichloro, c2h5sicl3
trichloro(ethyl)silane
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2041

RTECSNumber : VV4200000

TSCA : Yes

Documents