Please login to your online account to display your discounted pricing

Ethyltrichlorosilane, 99%, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging Glass Bottle

Manufacturer: fisher scientific acros  313511000

 View more versions of this product

Catalog No. AC313511000

  • / Each
 

Description & Specifications

Chemical Identifiers

Density 1.2380g/mL
Infrared Spectrum Authentic
Packaging Glass Bottle
Refractive Index 1.4240 to 1.4260 (20°C, 589nm)
Color Clear colorless to light yellow
Quantity 100mL
Chemical Name or Material Ethyltrichlorosilane
Name Note 95%
CAS 115-21-9
Physical Form Clear colorless to light yellow liquid
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C2H5Cl3Si
Linear Formula H5C2SiCl3
Formula Weight 163.51g/mol
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Reacts violently with water.
Contact with water liberates toxic gas.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Store in a dry place. Store in a closed container.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECS Number VV4200000
EINECS Number 204-072-6
TSCA TSCA

Specifications

Chemical Name or Material Ethyltrichlorosilane
Name Note 95%
CAS 115-21-9
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C2H5Cl3Si
Linear Formula H5C2SiCl3
Formula Weight 163.51g/mol
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Store in a dry place. Store in a closed container.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Reacts violently with water.
Contact with water liberates toxic gas.

SDS & Safety Documents