Please login to your online account to display your discounted pricing

(Ethylthio)trimethylsilane, 90%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (Ethylthio)trimethylsilane
Name Note 90%
CAS 5573-62-6
CAS Min % 88.0
CAS Max % 100.0
Purity Grade Technical Grade
Assay ≥88%
Assay Percent Range 90%
Molecular Formula C5H14SSi
Linear Formula CH5SSi(CH3)3
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC345000100

  SDS
thermo fisher scientific
345000100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC345000010

  SDS
thermo fisher scientific
345000010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥88%
Purity Grade Technical Grade
Physical Form Liquid
Formula Weight 134.3
Quantity 10g
Color Clear colorless
TSCA TSCA
Density 0.83g/mL
CAS Min % 88.0
CAS Max % 100.0
Refractive Index 1.4470 to 1.4510 (20°C, 589nm)
MDL Number MFCD00042890
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material (Ethylthio)trimethylsilane
Name Note 90%
Assay Percent Range 90%
Molecular Formula C5H14SSi
Linear Formula CH5SSi(CH3)3
CAS 5573-62-6
Infrared Spectrum Authentic
Packaging Glass bottle
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Highly flammable liquid and vapor.