Please login to your online account to display your discounted pricing

Alfa Aesar™ (Ethylthio)acetone, 95%

Chemical Identifiers

Chemical Name or Material (Ethylthio)acetone
CAS 20996-62-7
Molecular Formula C5H10OS
Formula Weight 118.2
MDL Number MFCD00026976
UN Number UN1993
Beilstein 1740830
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0414206

 
alfa aesar™
L0414206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0414214

 
alfa aesar™
L0414214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 118.2
CAS 20996-62-7
Chemical Name or Material (Ethylthio)acetone
Assay 95%
Molecular Formula C5H10OS
MDL Number MFCD00026976
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
UN Number UN1993
Beilstein 1740830
Health Hazard 1 Warning
Flash Point 52°C (125°F)
Boiling Point 170° to 172°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapor.