Please login to your online account to display your discounted pricing

Ethylmalonic Acid 97%, ACROS Organics™

$83.91 - $83.91

Chemical Identifiers

Chemical Name or Material Ethylmalonic acid
Name Note 97%
CAS 601-75-2
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H8O4
Linear Formula HO2CCH(C2H5)CO2H
Formula Weight 132.12
MDL Number MFCD00002668
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC146810100

  SDS
thermo fisher scientific
146810100
Glass bottle 10g Each for $83.91
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 132.12
Quantity 10g
Color White to faintly pink
Beilstein 02, IV, 1952
EINECS Number 210-007-2
Melting Point 109° to 114°C
Infrared Spectrum Authentic
Packaging Glass bottle
CAS 601-75-2
Linear Formula HO2CCH(C2H5)CO2H
Molecular Formula C5H8O4
Name Note 97%
Assay Percent Range 97%
Chemical Name or Material Ethylmalonic acid
MDL Number MFCD00002668