missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethylmagnesium chloride, 2.7M (25 wt.%) solution in THF, AcroSeal™, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific AC252570024

 View more versions of this product

Catalog No. AC252570024



Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Ethylmagnesium chloride
1.0000g/mL
C2H5ClMg
MFCD00000472
chloro ethyl magnesium, chloroethyl-magnesiu, chloroethylmagnesium, ethyl magnesium chloride, ethyl magnesiumchloride, ethylmagnesium chloride, ethylmagnesium chloride solution, ethylmagnesiumchloride, etmgcl, magnesium, chloroethyl
Solubility in water: reacts
1
magnesium;ethane;chloride
75432
Liquid
2386-64-3,109-99-9
Brown
C2H5MgCl
AcroSeal™ Glass Bottle
2.4L
YCCXQARVHOPWFJ-UHFFFAOYSA-M
C[CH2-].[Mg+2].[Cl-]
88.83
88.83
24 to 26%

Chemical Identifiers

2386-64-3
88.83
YCCXQARVHOPWFJ-UHFFFAOYSA-M
75432
C[CH2-].[Mg+2].[Cl-]
C2H5ClMg
MFCD00000472
chloro ethyl magnesium, chloroethyl-magnesiu, chloroethylmagnesium, ethyl magnesium chloride, ethyl magnesiumchloride, ethylmagnesium chloride, ethylmagnesium chloride solution, ethylmagnesiumchloride, etmgcl, magnesium, chloroethyl
magnesium;ethane;chloride
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
In contact with water releases flammable gases which may ignite spontaneously.
Highly flammable liquid and vapor.
Reacts violently w

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture

GHS Signal Word: Danger

EINECSNumber : 219-206-9

TSCA : TSCA

SDS Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only