Learn More
Ethylmagnesium chloride, 2.7M (25 wt.%) solution in THF, AcroSeal™, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific 252578000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Ethylmagnesium chloride | |
1.0000g/mL | |
C2H5ClMg | |
MFCD00000472 | |
chloro ethyl magnesium, chloroethyl-magnesiu, chloroethylmagnesium, ethyl magnesium chloride, ethyl magnesiumchloride, ethylmagnesium chloride, ethylmagnesium chloride solution, ethylmagnesiumchloride, etmgcl, magnesium, chloroethyl | |
Solubility in water: reacts | |
1 | |
magnesium;ethane;chloride | |
75432 | |
Liquid |
2386-64-3,109-99-9 | |
Brown | |
C2H5MgCl | |
AcroSeal™ Glass Bottle | |
800mL | |
YCCXQARVHOPWFJ-UHFFFAOYSA-M | |
C[CH2-].[Mg+2].[Cl-] | |
88.83 | |
88.83 | |
24 to 26% |
Chemical Identifiers
2386-64-3 | |
88.83 | |
YCCXQARVHOPWFJ-UHFFFAOYSA-M | |
75432 | |
C[CH2-].[Mg+2].[Cl-] |
C2H5ClMg | |
MFCD00000472 | |
chloro ethyl magnesium, chloroethyl-magnesiu, chloroethylmagnesium, ethyl magnesium chloride, ethyl magnesiumchloride, ethylmagnesium chloride, ethylmagnesium chloride solution, ethylmagnesiumchloride, etmgcl, magnesium, chloroethyl | |
magnesium;ethane;chloride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
In contact with water releases flammable gases which may ignite spontaneously.
Highly flammable liquid and vapor.
Reacts violently w
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture
GHS Signal Word: Danger
EINECSNumber : 219-206-9
TSCA : TSCA
RUO – Research Use Only