missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethylmagnesium bromide, 3M in ether, Thermo Scientific Chemicals

Grignard reagent

Supplier:  Thermo Scientific Chemicals 087291JT

 View more versions of this product

Catalog No. AA87291JT



Description

Description

Ethylmagnesium bromide is used as grignard reagent. It is widely used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethylmagnesium bromide
-116.3°C
1.020 g/mL
Ether-like
C2H5MgBr
0.5 mol
Air and moisture sensitive; May form a precipitate
Reacts with water.
CC[Mg]Br
133.27
133.28
Liquid
925-90-6
Brown to Yellow
34.6°C
C2H5BrMg
MFCD00000043
UN3399
ethylmagnesium bromide, grignard reagent, magnesium, bromoethyl, magnesium, bromoethyl, bromoethylmagnesium, ethylmagnesium bromide solution, 1.0 m in thf, ethylmagnesium bromide solution, 1.0 m in thf, ethylmagnesium bromide solution, 3.0 m in diethyl ether, ethylmagnesiumbromid, ethylmagnesiumbromid, ethylmagnesium bromid
TWTWFMUQSOFTRN-UHFFFAOYSA-M
magnesium;ethane;bromide
101914
3M in ether

Chemical Identifiers

925-90-6
133.27
TWTWFMUQSOFTRN-UHFFFAOYSA-M
101914
CC[Mg]Br
C2H5BrMg
MFCD00000043
ethylmagnesium bromide, grignard reagent, magnesium, bromoethyl, magnesium, bromoethyl, bromoethylmagnesium, ethylmagnesium bromide solution, 1.0 m in thf, ethylmagnesium bromide solution, 1.0 m in thf, ethylmagnesium bromide solution, 3.0 m in diethyl ether, ethylmagnesiumbromid, ethylmagnesiumbromid, ethylmagnesium bromid
magnesium;ethane;bromide
Safety and Handling

Safety and Handling

GHS H Statement
H224-H260-H314-H336
Extremely flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.

P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H224-H260-H302-H314-H336

DOTInformation : Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE

EINECSNumber : 213-127-3

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only