Please login to your online account to display your discounted pricing

Ethylmagnesium bromide, 0.9M in tert-Butyl methyl ether, AcroSeal™, ACROS Organics™


Quantity 800mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC347288000

 View more versions of this product

Catalog No. AC347288000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 133.27
Quantity 800mL
Color Dark brown
Fieser 02,205; 02,205
Packaging AcroSeal™ Glass Bottle
Density 0.8400g/mL
Chemical Name or Material Ethylmagnesium bromide
Name Note 0.9M Solution in tert-Butyl Methyl Ether
Molecular Formula C2H5BrMg
Concentration 13 to 16%
Linear Formula C2H5MgBr
MDL Number MFCD00000043
CAS 925-90-6
Health Hazard 1 Danger
Additional Information product may develop some turbidity or precipitate
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Highly flammable liquid and vapor.
Reacts violently with water.
CAS 1634-04-4
Health Hazard 3 GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Ethylmagnesium bromide
Name Note 0.9M Solution in tert-Butyl Methyl Ether
CAS 925-90-6
CAS 1634-04-4
Molecular Formula C2H5BrMg
Linear Formula C2H5MgBr
Formula Weight 133.27
MDL Number MFCD00000043
Accessories

Danger

GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Highly flammable liquid and vapor.
Reacts violently with water.

SDS & Safety Documents