missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethylmagnesium bromide, 0.9M in tert-Butyl methyl ether, AcroSeal™, ACROS Organics™

Chemical Identifiers

CAS 925-90-6,1634-04-4
Molecular Formula C2H5BrMg
Linear Formula C2H5MgBr
Formula Weight 133.27
MDL Number MFCD00000043
Chemical Name or Material Ethylmagnesium bromide, 0.9M Solution in tert-Butyl Methyl Ether
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC347281000 SDS Thermo Fisher Scientific
AC347281000
AcroSeal™ Glass Bottle in Tin Can 100mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC347288000 SDS Thermo Fisher Scientific
AC347288000
AcroSeal™ Glass Bottle 800mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

925-90-6,1634-04-4
C2H5MgBr
MFCD00000043
C2H5BrMg
133.27
Ethylmagnesium bromide, 0.9M Solution in tert-Butyl Methyl Ether

Specifications

925-90-6,1634-04-4
Liquid
13 to 16%
133.27
MFCD00000043
AcroSeal™ Glass Bottle in Tin Can
100mL
product may develop some turbidity or precipitate
C2H5BrMg
C2H5MgBr
0.8400g/mL
02,205; 02,205
Dark brown
Ethylmagnesium bromide, 0.9M Solution in tert-Butyl Methyl Ether

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Highly flammable liquid and vapor.
Reacts violently with water.

GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger