Please login to your online account to display your discounted pricing

Ethylidene diacetate, 99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  410131000

 View more versions of this product

Catalog No. AC410131000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5% (GC)
Physical Form Liquid
Formula Weight 146.14
Quantity 100g
Color Clear colorless to light yellow
Chemical Name or Material Ethylidene diacetate
Packaging Glass bottle
Molecular Formula C6H10O4
CAS 542-10-9
Synonym 1, 1-Ethanediol diacetate
Infrared Spectrum Authentic
Assay Percent Range 99%
Refractive Index 1.4000 to 1.4020 (20°C, 589nm)
Linear Formula CH3CH(OCOCH3)2

Specifications

Chemical Name or Material Ethylidene diacetate
Assay ≥98.5% (GC)
Assay Percent Range 99%
Molecular Formula C6H10O4
Linear Formula CH3CH(OCOCH3)2
Formula Weight 146.14
Synonym 1, 1-Ethanediol diacetate