Please login to your online account to display your discounted pricing

Ethylidene diacetate, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethylidene diacetate
Assay ≥98.5% (GC)
Assay Percent Range 99%
Molecular Formula C6H10O4
Linear Formula CH3CH(OCOCH3)2
Formula Weight 146.14
Synonym 1, 1-Ethanediol diacetate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC410130250

  SDS
fisher scientific acros
410130250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC410131000

  SDS
thermo fisher scientific
410131000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC410135000

  SDS
thermo fisher scientific
AC410135000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5% (GC)
Formula Weight 146.14
Physical Form Liquid
Quantity 25g
Color Clear colorless to light yellow
Refractive Index 1.4000 to 1.4020 (20°C, 589nm)
Linear Formula CH3CH(OCOCH3)2
Chemical Name or Material Ethylidene diacetate
Infrared Spectrum Authentic
Synonym 1, 1-Ethanediol diacetate
Packaging Glass bottle
Molecular Formula C6H10O4
CAS 542-10-9
Assay Percent Range 99%