Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethylhydrocupreine hydrochloride, 97%

$46.80 - $126.22

Chemical Identifiers

Chemical Name or Material Ethylhydrocupreine hydrochloride
CAS 3413-58-9
Molecular Formula C21H26N2O2·HCl
Formula Weight 374.91
MDL Number MFCD00135594
Synonym Optochin hydrochloride; Numoquin hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB21627MC

 
alfa aesar™
B21627MC
100mg Each for $46.80

AAB21627ME

 
alfa aesar™
B21627ME
500mg Each for $126.22
Description & Specifications

Specifications

Formula Weight 374.91
Chemical Name or Material Ethylhydrocupreine hydrochloride
Assay 97%
Molecular Formula C21H26N2O2·HCl
MDL Number MFCD00135594
CAS 3413-58-9
Synonym Optochin hydrochloride; Numoquin hydrochloride
Melting Point ≈240°C (dec.)
Quantity 100mg
Recommended Storage 2° to 8°C
RTECS Number MW6300000
Safety and Handling

2° to 8°C