Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethylferrocene, 98%

$26.16 - $258.59

Chemical Identifiers

Chemical Name or Material Ethylferrocene
CAS 1273-89-8
Formula Weight 214.09
MDL Number MFCD00059071
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3925302

  SDS
alfa aesar™
3925302
0.5g Each for $26.16

AA3925304

  SDS
alfa aesar™
3925304
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3925309

  SDS
alfa aesar™
3925309
10g Each for $258.59
Description & Specifications

Specifications

Formula Weight 214.09
Chemical Name or Material Ethylferrocene
Density 1.256g/mL
TSCA TSCA
Physical Form Liquid
Assay 98%
Refractive Index 1.6010
MDL Number MFCD00059071
CAS 1273-89-8
Boiling Point 264°C
Quantity 0.5g