Please login to your online account to display your discounted pricing

ethylevalerate, 99%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC410475000

 View more versions of this product

Catalog No. AC410475000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 130.19
Quantity 500g
Color Clear colorless
Density 0.8700g/mL
Merck Index 15, 10090
Refractive Index 1.3990 to 1.4010 (20°C, 589nm)
Packaging Glass bottle
Linear Formula CH3(CH2)3COOC2H5
Molecular Formula C7H14O2
Assay Percent Range 99%
CAS 539-82-2
Chemical Name or Material Ethyl valerate
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00009479
Infrared Spectrum Authentic
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.

Specifications

Chemical Name or Material Ethyl valerate
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H14O2
Linear Formula CH3(CH2)3COOC2H5
Formula Weight 130.19
MDL Number MFCD00009479
Accessories

Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapor.

SDS & Safety Documents