Please login to your online account to display your discounted pricing

ethyletrans-β-methylcinnamate, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC428000050

 View more versions of this product

Catalog No. AC428000050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 190.24
Quantity 5g
Density 1.0420g/mL
Molecular Formula C12H14O2
Chemical Name or Material Ethyl trans-β-methylcinnamate
Packaging Glass bottle
Assay Percent Range 97%
Linear Formula C6H5C(CH3)=CHCO2C2H5
MDL Number MFCD00053762
CAS 1504-72-9

Specifications

Chemical Name or Material Ethyl trans-β-methylcinnamate
Assay Percent Range 97%
Molecular Formula C12H14O2
Linear Formula C6H5C(CH3)=CHCO2C2H5
Formula Weight 190.24
MDL Number MFCD00053762