missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
ethylethiomorpholine-3-carboxylate hydrochloride, 90%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC27673DE
Specifications
ethylethiomorpholine-3-carboxylate hydrochloride | |
89.5 | |
90% | |
C7H14ClNO2S | |
5g | |
QQRCEZBURAQLBQ-UHFFFAOYSA-N | |
ethyl thiomorpholine-3-carboxylate;hydrochloride | |
17433447 | |
90% |
159381-07-4 | |
100.0 | |
Amber Glass Bottle | |
MFCD00107968 | |
ethyl thiomorpholine-3-carboxylate hydrochloride, thiomorpholine-3-carboxylic acid ethyl ester hydrochloride, thiomorpholine-3-carboxylic acid ethyl ester hcl, thiomorpholine-3-carboxylicacidethylesterhydrochloride, ethyl thiomorpholine-3-carboxylate hcl, ethylthiomorpholine-3-carboxylatehydrochloride, 3-ethoxycarbonyl thiomorpholine hydrochloride, ethyl 1,4-thiazinane-3-carboxylate hydrochloride, ethyl 1,4-thiazaperhydroine-3-carboxylate, chloride, thiomorpholine-3-carboxylic acid ethyl esterhydrochloride | |
CCOC(=O)C1CSCCN1.Cl | |
211.704 | |
211.71 |
Chemical Identifiers
159381-07-4 | |
211.704 | |
QQRCEZBURAQLBQ-UHFFFAOYSA-N | |
17433447 | |
CCOC(=O)C1CSCCN1.Cl |
C7H14ClNO2S | |
MFCD00107968 | |
ethyl thiomorpholine-3-carboxylate hydrochloride, thiomorpholine-3-carboxylic acid ethyl ester hydrochloride, thiomorpholine-3-carboxylic acid ethyl ester hcl, thiomorpholine-3-carboxylicacidethylesterhydrochloride, ethyl thiomorpholine-3-carboxylate hcl, ethylthiomorpholine-3-carboxylatehydrochloride, 3-ethoxycarbonyl thiomorpholine hydrochloride, ethyl 1,4-thiazinane-3-carboxylate hydrochloride, ethyl 1,4-thiazaperhydroine-3-carboxylate, chloride, thiomorpholine-3-carboxylic acid ethyl esterhydrochloride | |
ethyl thiomorpholine-3-carboxylate;hydrochloride |