Please login to your online account to display your discounted pricing

ethylestearate, mixture 1:1 with ethylepalmitate, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  327650050

 View more versions of this product

Catalog No. AC327650050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≈50% ethyl stearate, ≈50% ethyl palmitate
Physical Form Crystalline Mass
Formula Weight 312.54
Quantity 5g
Color White
Melting Point 22° to 26°C
Additional Information this product may appear as a liquid
Chemical Name or Material ethylestearate
Molecular Formula C20H40O2
MDL Number MFCD00009006
CAS 111-61-5
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material ethylestearate
Assay ≈50% ethyl stearate, ≈50% ethyl palmitate
Molecular Formula C20H40O2
Formula Weight 312.54
MDL Number MFCD00009006