missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ethylestearate, mixture 1:1 with ethylepalmitate, ACROS Organics™

Chemical Identifiers

Molecular Formula C20H40O2
Formula Weight 312.54
MDL Number MFCD00009006
Chemical Name or Material ethylestearate
Assay ≈50% ethyl stearate, ≈50% ethyl palmitate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC327650050 SDS Thermo Fisher Scientific
327650050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

C20H40O2
MFCD00009006
≈50% ethyl stearate, ≈50% ethyl palmitate
312.54
ethylestearate

Specifications

this product may appear as a liquid
22° to 26°C
111-61-5
Glass bottle
C20H40O2
MFCD00009006
≈50% ethyl stearate, ≈50% ethyl palmitate
White
5g
Authentic
Crystalline Mass
312.54
ethylestearate