missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl propiolate, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC118901000

 View more versions of this product

Catalog No. AC118901000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Ethyl propiolate
23°C
Glass bottle
0.968
Colorless to Yellow
623-47-2
C5H6O2
MFCD00009184
15, 3898
Solubility in water: miscible with water. Other solubilities: soluble in most organic solvents
CCOC(=O)C#C
98.1
CHEBI:51740
Liquid
0.9680g/mL
Authentic
1.4110 to 1.413
120°C
100g
98.5% min. (GC)
HC∽CCO2C2H5
02, 477
2-propynoic acid, ethyl ester, ethoxycarbonyl acetylene, ethyl 2-propynoate, ethyl acetylenecarboxylate, ethyl propiolate, ethyl propynoate, propiolic acid ethyl ester, propiolic acid, ethyl ester, propynoic acid ethyl ester, unii-w235g5u52s
FMVJYQGSRWVMQV-UHFFFAOYSA-N
ethyl prop-2-ynoate
12182
98.1
99%

Chemical Identifiers

623-47-2
98.1
FMVJYQGSRWVMQV-UHFFFAOYSA-N
12182
ethyl prop-2-ynoate
C5H6O2
MFCD00009184
2-propynoic acid, ethyl ester, ethoxycarbonyl acetylene, ethyl 2-propynoate, ethyl acetylenecarboxylate, ethyl propiolate, ethyl propynoate, propiolic acid ethyl ester, propiolic acid, ethyl ester, propynoic acid ethyl ester, unii-w235g5u52s
CHEBI:51740
CCOC(=O)C#C
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 210-795-8

SDS Documents